Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-33C11
Entrez Gene ID
190065
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_Y49C4A.9
Chromosome Location
Chr.V: 710453 - 712982 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070902.2 NP_503303.1 3 - 4 1273 134

View Full Product Details