Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-25
Entrez Gene ID
191943
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_C05E4.10
Chromosome Location
Chr.V: 734420 - 736271 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070907.1 NP_503308.1 4 - 5 439 158

View Full Product Details