Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srab-16
Entrez Gene ID
187865
Gene Name
protein-coding
Gene Aliases
CELE_R13D11.6
Chromosome Location
Chr.V: 782732 - 784763 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070924.3 NP_503325.2 5 - 6 843 83

View Full Product Details