Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-12
Entrez Gene ID
187201
Gene Name
protein-coding
Gene Aliases
CELE_K09C6.4
Chromosome Location
Chr.V: 849812 - 851332 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313217.1 NP_001300146.1 3 - 4 511 73
NM_070940.2 NP_503341.2 4 - 5 619 73

View Full Product Details