Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-23
Entrez Gene ID
191941
Gene Name
protein-coding
Gene Aliases
CELE_R09B5.7
Chromosome Location
Chr.V: 1451634 - 1454340 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028901.3 NP_001024072.1 3 - 4 448 94
NM_001028902.1 NP_001024073.1 3 - 4 448 94

View Full Product Details