Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
F53E2.2
Entrez Gene ID
41657006
Gene Name
Actin-depolymerizing factor 1, isoforms a/b
Gene Aliases
CELE_F53E2.2
Chromosome Location
Chr.V: 1475086 - 1479577 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071024.8 NP_503425.2 2 - 3 150 61

View Full Product Details