Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-41
Entrez Gene ID
184561
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_F16B4.10
Chromosome Location
Chr.V: 1612803 - 1614826 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071055.2 NP_503456.2 3 - 4 622 163

View Full Product Details