Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-90
Entrez Gene ID
189966
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y46H3C.3
Chromosome Location
Chr.V: 1702051 - 1704053 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071085.1 NP_503486.1 3 - 4 675 132

View Full Product Details