Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gly-7
Entrez Gene ID
178661
Gene Name
protein-coding
Gene Aliases
CELE_Y46H3A.6
Chromosome Location
Chr.V: 1815851 - 1821323 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313323.4 NP_001300252.1 3 - 4 1536 63
NM_001313324.3 NP_001300253.1 2 - 3 849 63
NM_071111.6 NP_503512.1 5 - 6 1698 63
GenBank mRNA AF031841.1 - 5 - 6 1705 63

View Full Product Details