Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
alh-5
Entrez Gene ID
178680
Gene Name
Aldehyde dehydrogenase
Gene Aliases
CELE_T08B1.3
Chromosome Location
Chr.V: 1996100 - 1999581 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071144.6 NP_503545.1 3 - 4 784 74

View Full Product Details