Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-86
Entrez Gene ID
178685
Gene Name
protein-coding
Gene Aliases
CELE_Y40B10A.8
Chromosome Location
Chr.V: 2061106 - 2066850 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071160.9 NP_503561.2 4 - 5 889 64

View Full Product Details