Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-56
Entrez Gene ID
178699
Gene Name
protein-coding
Gene Aliases
CELE_F44C8.6
Chromosome Location
Chr.V: 2219004 - 2221305 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038398.3 NP_001033487.1 1 - 2 232 141
NR_133578.1 1 - 2 101 141

View Full Product Details