Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-62
Entrez Gene ID
182844
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_C24B9.12
Chromosome Location
Chr.V: 2740444 - 2742398 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380620.1 NP_001367562.1 2 - 3 412 92

View Full Product Details