Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-5
Entrez Gene ID
183701
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_C50H11.14
Chromosome Location
Chr.V: 3079449 - 3080892 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071440.2 NP_503841.2 1 - 2 157 107

View Full Product Details