Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-36
Entrez Gene ID
182098
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C02A12.6
Chromosome Location
Chr.V: 3489228 - 3490535 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071573.5 NP_503974.2 1 - 2 113 129

View Full Product Details