Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-64
Entrez Gene ID
178774
Gene Name
protein-coding
Gene Aliases
CELE_C07G3.9
Chromosome Location
Chr.V: 3503080 - 3506352 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071577.9 NP_503978.1 2 - 3 217 77

View Full Product Details