Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-64
Entrez Gene ID
178774
Gene Name
protein-coding
Gene Aliases
CELE_C07G3.9
Chromosome Location
Chr.V: 3503080 - 3506352 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322526.3 NP_001309621.1 3 - 4 396 115
NM_071577.9 NP_503978.1 5 - 6 636 115

View Full Product Details