Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-23
Entrez Gene ID
191606
Gene Name
protein-coding
Gene Aliases
CELE_F59B1.9
Chromosome Location
Chr.V: 3651656 - 3659572 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330798.2 NP_001317822.1 2 - 3 246 73
NM_071623.5 NP_504024.2 2 - 3 313 73
GenBank mRNA AY519853.1 - 2 - 3 249 73

View Full Product Details