Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-67
Entrez Gene ID
187098
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_K07C6.10
Chromosome Location
Chr.V: 3916780 - 3918538 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071683.5 NP_504084.3 3 - 4 466 116

View Full Product Details