Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-34A5
Entrez Gene ID
181847
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_B0213.10
Chromosome Location
Chr.V: 3955701 - 3957715 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071698.3 NP_504099.1 3 - 4 799 91

View Full Product Details