Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srt-13
Entrez Gene ID
191087
Gene Name
protein-coding
Gene Aliases
CELE_ZC142.1
Chromosome Location
Chr.V: 4247955 - 4250147 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071772.3 NP_504173.2 1 - 2 152 110

View Full Product Details