Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-14
Entrez Gene ID
186767
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_H23N18.2
Chromosome Location
Chr.V: 4906253 - 4908304 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071915.3 NP_504316.1 2 - 3 239 140

View Full Product Details