Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- srh-20
- Entrez Gene ID
-
191847
- Gene Name
- Serpentine Receptor, class H
- Gene Aliases
-
CELE_C02E7.3
- Chromosome Location
-
Chr.V: 4923983 - 4926221 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071923.4 |
NP_504324.1 |
1 - 2 |
379 |
116 |
- Gene Symbol
- srh-22
- Entrez Gene ID
-
191848
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C02E7.4
- Chromosome Location
-
Chr.V: 4922324 - 4924594 on Build WBcel235
View Full Product Details