Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-13
Entrez Gene ID
182574
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C13D9.6
Chromosome Location
Chr.V: 4981809 - 4983191 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071939.1 NP_504340.1 2 - 3 335 160

View Full Product Details