Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srr-4
Entrez Gene ID
187292
Gene Name
protein-coding
Gene Aliases
CELE_K11D12.3
Chromosome Location
Chr.V: 5019018 - 5022462 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071946.9 NP_504347.2 2 - 3 198 100

View Full Product Details