Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-63
Entrez Gene ID
182223
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_C04F5.7
Chromosome Location
Chr.V: 5108180 - 5110791 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071968.6 NP_504369.1 3 - 4 708 71

View Full Product Details