Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-51
Entrez Gene ID
178905
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_C03A7.11
Chromosome Location
Chr.V: 5175020 - 5177309 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071987.6 NP_504388.2 5 - 6 1257 146

View Full Product Details