Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-25
Entrez Gene ID
188326
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_T09D3.5
Chromosome Location
Chr.V: 5342094 - 5345519 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072023.4 NP_504424.3 6 - 7 1137 100

View Full Product Details