Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acy-4
Entrez Gene ID
178949
Gene Name
Adenylate cyclase
Gene Aliases
CELE_T01C2.1
Chromosome Location
Chr.V: 5624727 - 5630892 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072085.6 NP_504486.4 7 - 8 744 96

View Full Product Details