Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-16
Entrez Gene ID
191805
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C04E6.9
Chromosome Location
Chr.V: 5898538 - 5900040 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072135.1 NP_504536.1 2 - 3 289 116

View Full Product Details