Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sago-1
Entrez Gene ID
179016
Gene Name
Piwi-like protein
Gene Aliases
CELE_K12B6.1
Chromosome Location
Chr.V: 6303714 - 6308337 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072209.4 NP_504610.1 1 - 2 248 74

View Full Product Details