Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fut-2
Entrez Gene ID
184050
Gene Name
Galactoside 2-alpha-L-fucosyltransferase
Gene Aliases
CELE_EGAP9.2
Chromosome Location
Chr.V: 6573939 - 6575890 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072270.2 NP_504671.1 5 - 6 587 101

View Full Product Details