Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pmp-5
Entrez Gene ID
179056
Gene Name
protein-coding
Gene Aliases
CELE_T10H9.5
Chromosome Location
Chr.V: 6662182 - 6666524 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028960.5 NP_001024131.1 9 - 10 1169 70
NM_072288.6 NP_504689.1 8 - 9 1032 70

View Full Product Details