Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-68
Entrez Gene ID
179080
Gene Name
protein-coding
Gene Aliases
CELE_K11C4.5
Chromosome Location
Chr.V: 6901681 - 6929038 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356673.4 NP_001343696.1 44 - 44 14892 122
NM_001356674.3 NP_001343811.1 44 - 44 14895 122
NM_001356675.3 NP_001343689.1 45 - 45 14934 122
NM_001356676.3 NP_001343809.1 45 - 45 14937 122
NM_001356677.4 NP_001343695.1 46 - 46 15240 122
NM_001356678.4 NP_001343810.1 46 - 46 15243 122
NM_001356679.3 NP_001343688.1 47 - 48 15371 122
NM_001356680.4 NP_001343808.1 47 - 47 15285 122
NM_001356681.3 NP_001343815.1 44 - 45 14886 122
NM_001356682.3 NP_001343819.1 44 - 45 14889 122
NM_001356683.3 NP_001343813.1 45 - 46 14928 122
NM_001356684.3 NP_001343817.1 45 - 46 14931 122
NM_001356685.3 NP_001343814.1 46 - 47 15234 122
NM_001356686.3 NP_001343818.1 46 - 47 15237 122
NM_001356687.3 NP_001343812.1 47 - 48 15276 122
NM_001356688.3 NP_001343816.1 47 - 48 15279 122

View Full Product Details