Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-36
Entrez Gene ID
187946
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T01C4.6
Chromosome Location
Chr.V: 7129707 - 7131074 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072406.2 NP_504807.2 5 - 6 646 110

View Full Product Details