Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gei-6
Entrez Gene ID
179114
Gene Name
GEX Interacting protein
Gene Aliases
CELE_C05C8.4
Chromosome Location
Chr.V: 7226376 - 7230848 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072435.7 NP_504836.1 1 - 2 79 77

View Full Product Details