Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-35A2
Entrez Gene ID
182177
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_C03G6.15
Chromosome Location
Chr.V: 7362319 - 7364236 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072479.6 NP_504880.2 5 - 6 933 109

View Full Product Details