Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-6
Entrez Gene ID
187860
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_R13D7.10
Chromosome Location
Chr.V: 7407547 - 7408814 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072496.1 NP_504897.1 3 - 4 717 104

View Full Product Details