Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-10
Entrez Gene ID
182769
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C18B10.2
Chromosome Location
Chr.V: 7491875 - 7493024 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322571.1 NP_001309476.1 1 - 2 140 154

View Full Product Details