Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-11
Entrez Gene ID
182768
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C18B10.1
Chromosome Location
Chr.V: 7493880 - 7495289 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072528.4 NP_504929.1 4 - 5 670 69

View Full Product Details