Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
folt-3
Entrez Gene ID
4363081
Gene Name
Folate-like transporter 3
Gene Aliases
CELE_C50E3.16
Chromosome Location
Chr.V: 7603735 - 7609946 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047632.3 NP_001041097.2 5 - 6 514 136

View Full Product Details