Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-29
Entrez Gene ID
191812
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_F07C4.3
Chromosome Location
Chr.V: 7635769 - 7636882 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072574.1 NP_504975.1 2 - 3 663 114

View Full Product Details