Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-6
Entrez Gene ID
190818
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y97E10B.3
Chromosome Location
Chr.V: 7912276 - 7928470 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072636.2 NP_505037.1 12 - 13 1459 135

View Full Product Details