Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cogc-6
Entrez Gene ID
179201
Gene Name
Conserved oligomeric Golgi complex subunit 6
Gene Aliases
CELE_K07C11.9
Chromosome Location
Chr.V: 8218267 - 8221316 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072717.5 NP_505118.1 5 - 6 1709 122

View Full Product Details