Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
otpl-7
Entrez Gene ID
185808
Gene Name
OToPetrin-Like
Gene Aliases
CELE_F45F2.7
Chromosome Location
Chr.V: 8521503 - 8524678 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072793.4 NP_505194.2 5 - 6 775 149

View Full Product Details