Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hum-2
Entrez Gene ID
179321
Gene Name
protein-coding
Gene Aliases
CELE_F36D4.3
Chromosome Location
Chr.V: 9386175 - 9400067 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330875.4 NP_001317742.1 7 - 8 3011 97
NM_001330876.2 NP_001317741.1 7 - 8 3011 97
NM_001373073.2 NP_001359965.1 8 - 9 2938 97
NM_001380727.2 NP_001367720.1 7 - 8 3011 97

View Full Product Details