Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
his-41
Entrez Gene ID
179340
Gene Name
Probable histone H2B 3
Gene Aliases
CELE_C50F4.5
Chromosome Location
Chr.V: 9545522 - 9549739 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073063.5 NP_505464.1 1 - 2 192 84

View Full Product Details