Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ehbp-1
Entrez Gene ID
179343
Gene Name
EH (Eps-15-homology) domain Binding Protein family
Gene Aliases
CELE_F25B3.1
Chromosome Location
Chr.V: 9550644 - 9556033 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073067.4 NP_505468.1 1 - 2 73 89

View Full Product Details