Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-31
Entrez Gene ID
183647
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_C50C10.4
Chromosome Location
Chr.V: 9816690 - 9817862 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073133.1 NP_505534.1 2 - 3 451 133

View Full Product Details