Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-46
Entrez Gene ID
179384
Gene Name
Transthyretin-like protein 46
Gene Aliases
CELE_T07C12.7
Chromosome Location
Chr.V: 9951233 - 9952183 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073160.6 NP_505561.1 1 - 2 152 81

View Full Product Details